Merck
Ca2+-H+ antiport activity in synaptic vesicles isolated from sheep brain cortex.
Neuroscience letters (1998-07-09)
P P Gonçalves, S M Meireles, C Gravato, M G Vale
ABSTRAKT
Synaptic vesicles isolated from sheep brain cortex exhibit an ATP-dependent Ca2+ accumulation that is inhibited by the protonophore uncoupler carbonyl cyanide m-chorophenylhydrazone (CCCP) and completely released by the Ca2+ ionophore ionomycin. This transport activity was sensitive to the V-type ATPase inhibitor, bafilomycin, but not to the P-type ATPase inhibitor, vanadate. We also observed that the proton gradient, established across the synaptic vesicle membranes in the presence of ATP, is partially dissipated by the addition of Ca2+ (100-860 microM) in correlation to an increase of ATP hydrolysis by the H+-pumping ATPase. In contrast, the activity of the H+-ATPase, measured under uncoupling conditions (presence of CCCP), appears to be unaltered by the calcium ion. The Ca2+-induced H+ release visualized by fluorescence quenching of acridine orange correlates well with the Ca2+ uptake determined isotopically. These results indicate that synaptic vesicles accumulate Ca2+, via a low affinity Ca2+-H+ antiport system energized by the protonmotive force originated from the H+-pumping ATPase activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
QuickLink DNA Ligation Kit, joins blunt-end and sticky-end DNA fragments
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder