Merck
INS(GFP/w) human embryonic stem cells facilitate isolation of in vitro derived insulin-producing cells.
Diabetologia (2011-11-29)
S J Micallef, X Li, J V Schiesser, C E Hirst, Q C Yu, S M Lim, M C Nostro, D A Elliott, F Sarangi, L C Harrison, G Keller, A G Elefanty, E G Stanley
ABSTRAKT
We aimed to generate human embryonic stem cell (hESC) reporter lines that would facilitate the characterisation of insulin-producing (INS⁺) cells derived in vitro. Homologous recombination was used to insert sequences encoding green fluorescent protein (GFP) into the INS locus, to create reporter cell lines enabling the prospective isolation of viable INS⁺ cells. Differentiation of INS(GFP/w) hESCs using published protocols demonstrated that all GFP⁺ cells co-produced insulin, confirming the fidelity of the reporter gene. INS-GFP⁺ cells often co-produced glucagon and somatostatin, confirming conclusions from previous studies that early hESC-derived insulin-producing cells were polyhormonal. INS(GFP/w) hESCs were used to develop a 96-well format spin embryoid body (EB) differentiation protocol that used the recombinant protein-based, fully defined medium, APEL. Like INS-GFP⁺ cells generated with other methods, those derived using the spin EB protocol expressed a suite of pancreatic-related transcription factor genes including ISL1, PAX6 and NKX2.2. However, in contrast with previous methods, the spin EB protocol yielded INS-GFP⁺ cells that also co-expressed the beta cell transcription factor gene, NKX6.1, and comprised a substantial proportion of monohormonal INS⁺ cells. INS(GFP/w) hESCs are a valuable tool for investigating the nature of early INS⁺ progenitors in beta cell ontogeny and will facilitate the development of novel protocols for generating INS⁺ cells from differentiating hESCs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L)-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder