Merck
Redirecting gene-modified T cells toward various cancer types using tagged antibodies.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-10-04)
Koji Tamada, Degui Geng, Yukimi Sakoda, Navneeta Bansal, Ratika Srivastava, Zhaoyang Li, Eduardo Davila
ABSTRAKT
To develop an adaptable gene-based vector that will confer immune cell specificity to various cancer types. Human and mouse T cells were genetically engineered to express a chimeric antigen receptor (CAR) that binds a fluorescein isothiocyanate (FITC) molecule, termed anti-FITC CAR T cells. Various antibodies (Ab) currently in clinical use including cetuximab (Ctx), trastuzumab (Her2), and rituximab (Rtx) were conjugated with FITC and tested for their ability to bind tumor cells, activate T cells, and induce antitumor effects in vitro and in vivo. Anti-FITC CAR T cells recognize various cancer types when bound with FITC-labeled Abs resulting in efficient target lysis, T-cell proliferation, and cytokine/chemokine production. The treatment of immunocompromised mice with human anti-FITC CAR T cells plus FITC-labeled cetuximab (FITC-Ctx) delayed the growth of colon cancer but unexpectedly led to the outgrowth of EGF receptor (EGFR)-negative tumor cells. On the other hand, in a human pancreatic cancer cell line with uniform EGFR expression, anti-FITC CAR T cells plus FITC-Ctx eradicated preestablished late-stage tumors. In immunocompetent mice, anti-FITC CAR T cells exhibited potent antitumor activity against syngeneic mouse breast cancer expressing Her2 and B-cell lymphoma expressing CD20 by combining with FITC-Her2 and FITC-Rtx, respectively. In addition, the activity of anti-FITC CAR T cells could be attenuated by subsequent injections of nonspecific FITC-IgG. These studies highlight an applicability of anti-tag CAR technology to treat patients with different types of cancers and a possibility to regulate CAR T-cell functions with competing FITC molecules.