Merck
Mutations in Fibronectin Cause a Subtype of Spondylometaphyseal Dysplasia with "Corner Fractures".
American journal of human genetics (2017-11-04)
Chae Syng Lee, He Fu, Nissan Baratang, Justine Rousseau, Heena Kumra, V Reid Sutton, Marcello Niceta, Andrea Ciolfi, Guilherme Yamamoto, Débora Bertola, Carlo L Marcelis, Dorien Lugtenberg, Andrea Bartuli, Choel Kim, Julie Hoover-Fong, Nara Sobreira, Richard Pauli, Carlos Bacino, Deborah Krakow, Jillian Parboosingh, Patrick Yap, Ariana Kariminejad, Marie T McDonald, Mariana I Aracena, Ekkehart Lausch, Sheila Unger, Andrea Superti-Furga, James T Lu, Dan H Cohn, Marco Tartaglia, Brendan H Lee, Dieter P Reinhardt, Philippe M Campeau
ABSTRAKT
Fibronectin is a master organizer of extracellular matrices (ECMs) and promotes the assembly of collagens, fibrillin-1, and other proteins. It is also known to play roles in skeletal tissues through its secretion by osteoblasts, chondrocytes, and mesenchymal cells. Spondylometaphyseal dysplasias (SMDs) comprise a diverse group of skeletal dysplasias and often manifest as short stature, growth-plate irregularities, and vertebral anomalies, such as scoliosis. By comparing the exomes of individuals with SMD with the radiographic appearance of "corner fractures" at metaphyses, we identified three individuals with fibronectin (FN1) variants affecting highly conserved residues. Furthermore, using matching tools and the SkelDys emailing list, we identified other individuals with de novo FN1 variants and a similar phenotype. The severe scoliosis in most individuals and rare developmental coxa vara distinguish individuals with FN1 mutations from those with classical Sutcliffe-type SMD. To study functional consequences of these FN1 mutations on the protein level, we introduced three disease-associated missense variants (p.Cys87Phe [c.260G>T], p.Tyr240Asp [c.718T>G], and p.Cys260Gly [c.778T>G]) into a recombinant secreted N-terminal 70 kDa fragment (rF70K) and the full-length fibronectin (rFN). The wild-type rF70K and rFN were secreted into the culture medium, whereas all mutant proteins were either not secreted or secreted at significantly lower amounts. Immunofluorescence analysis demonstrated increased intracellular retention of the mutant proteins. In summary, FN1 mutations that cause defective fibronectin secretion are found in SMD, and we thus provide additional evidence for a critical function of fibronectin in cartilage and bone.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fibronectin human, recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture