Merck
  • 5-Hydroxymethylcytosine (5hmC) and Ten-eleven translocation 1-3 (TET1-3) proteins in the dorsal root ganglia of mouse: Expression and dynamic regulation in neuropathic pain.
5-Hydroxymethylcytosine (5hmC) and Ten-eleven translocation 1-3 (TET1-3) proteins in the dorsal root ganglia of mouse: Expression and dynamic regulation in neuropathic pain.
Somatosensory & motor research (2017-03-10)
Alexander G Chamessian, Yawar J Qadri, Matthew Cummins, Michele Hendrickson, Temugin Berta, Thomas Buchheit, Thomas Van de Ven
ABSTRAKT
Epigenetic mechanisms are increasingly implicated in chronic pain pathology. In this study, we demonstrate that the novel epigenetic mark 5-hydroxymethylcytosine (5hmC) is present in dorsal root ganglia (DRG) neurons and glia, and its levels increase following nerve injury. Furthermore, we show that the 5hmC-generating Ten-eleven translocation 1-3 (TET1-3) proteins are expressed in a cell-type specific manner in the DRG, with Tet3 displaying differential upregulation after injury, suggesting a potential role in neuropathic pain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-S-100 (β-Subunit) antibody, Mouse monoclonal, clone SH-B4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Tet2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse