Merck
Influence of POLG on Radiosensitivity of Nasopharyngeal Carcinoma Cells.
Cancer biotherapy & radiopharmaceuticals (2018-05-16)
Ruiqing Chen, Zeng Wang, Ruilong Lan, Fei Huang, Jinrong Chen, Yuanteng Xu, Hengshan Zhang, Lurong Zhang
ABSTRAKT
There is a high incidence of nasopharyngeal carcinoma (NPC), malignant head and neck tumors, in southern China. Radioresistance is the main cause affecting the efficacy of NPC treatments. The POLG gene particularly plays an important role in radiation-induced damage repair. In this study, the authors established RNAi CNE-1 and CNE-2 knockdown in two NPC cell lines to observe whether this gene affects the radiosensitivity of NPC cells. Four short hairpin RNA (shRNA) expression plasmids targeting POLG gene were constructed and transfected into the NPC cell lines CNE-1 and CNE-2. Screening was performed to evaluate the stable expression of cloned cells, which were named CNE-1/POLG-shRNA1, CNE-1/POLG-shRNA2, CNE-2/POLG-shRNA1, and CNE-2/POLG-shRNA2. The negative controls CNE-1/Neg-shRNA and CNE-2/Neg-shRNA were additionally used. The MTT method, flow cytometry, clone formation analysis, cell migration, and other experimental methods were employed to verify changes in the radiosensitivity of the NPC cells. Fluorescent quantitative PCR and Western blot confirmed the downregulation of the PLOG gene through diminished PLOG messenger RNA and protein levels. Consequently, the authors report the stable knockdown of the POLG gene in an NPC model. Dose-dependent radiation exposure of POLG inhibited NPC cell growth and increased apoptosis compared with control cells (p < 0.01), as demonstrated through colony formation assay and flow cytometry. Functional assays indicated that knockdown of the POLG in CNE-1 and CNE-2 cells remarkably reduced cell viability and proliferation. Specifically, POLG knockdown led to G1 phase arrest and apoptosis. Overall, the authors conclude that POLG downregulation alters the radiosensitivity of NPC cells, indicating that the gene is likely involved in conferring the radiation response of the cells. In addition, findings in this study suggest a novel role for POLG as a potential predictive marker for NPC radiotherapy efficiency. POLG gene can be used as a potential clinical target to effectively improve the radiosensitivity of NPC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human POLG