Merck
  • Biotransformation and oxidative stress responses in rat hepatic cell-line (H4IIE) exposed to racemic ketoprofen (RS-KP) and its enantiomer, dexketoprofen (S(+)-KP).
Biotransformation and oxidative stress responses in rat hepatic cell-line (H4IIE) exposed to racemic ketoprofen (RS-KP) and its enantiomer, dexketoprofen (S(+)-KP).
Environmental toxicology and pharmacology (2018-04-08)
Elvira Mennillo, Åse Krøkje, Carlo Pretti, Valentina Meucci, Augustine Arukwe
ABSTRAKT
Pharmaceuticals such as racemate ketoprofen (RS-KP) and its enantiomer, dexketoprofen (S(+)-KP) are highly detectable non-steroidal anti-inflammatory drugs (NSAIDs) in the aquatic environment and therefore are designated as one of the most emerging groups of pollutants that can affect environmental and human health. The potential impact of these pharmaceuticals was assessed for the first time in vitro using a rat hepatocellular carcinoma cell line (H4IIE). Cells were exposed to low and high concentrations of these drugs. Cytotoxicity was determined by MTT reduction assay; CYP1A1 transcriptional and enzymatic levels together with canonical oxidative stress responsive markers (GPx, GR, GST and CAT) were also investigated. Cells exposed to RS-KP and S(+)-KP did not show cytotoxicity effect at the concentrations tested. However, this study highlighted differences between RS-KP and S(+)-KP in most of the evaluated markers, showing compound-, concentration- and time-specific effect patterns which suggest a potential stereo-selective toxicity of these drugs.