Merck
Cutting Edge: Mitochondrial Assembly of the NLRP3 Inflammasome Complex Is Initiated at Priming.
Journal of immunology (Baltimore, Md. : 1950) (2018-04-01)
Eric I Elliott, Alexis N Miller, Balaji Banoth, Shankar S Iyer, Aleksandr Stotland, Jerrold P Weiss, Roberta A Gottlieb, Fayyaz S Sutterwala, Suzanne L Cassel
ABSTRAKT
The NLRP3 inflammasome is activated in response to microbial and danger signals, resulting in caspase-1-dependent secretion of the proinflammatory cytokines IL-1β and IL-18. Canonical NLRP3 inflammasome activation is a two-step process requiring both priming and activation signals. During inflammasome activation, NLRP3 associates with mitochondria; however, the role for this interaction is unclear. In this article, we show that mouse NLRP3 and caspase-1 independently interact with the mitochondrial lipid cardiolipin, which is externalized to the outer mitochondrial membrane at priming in response to reactive oxygen species. An NLRP3 activation signal is then required for the calcium-dependent association of the adaptor molecule ASC with NLRP3 on the mitochondrial surface, resulting in inflammasome complex assembly and activation. These findings demonstrate a novel lipid interaction for caspase-1 and identify a role for mitochondria as supramolecular organizing centers in the assembly and activation of the NLRP3 inflammasome.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt, ≥95% (H-NMR), powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®