Merck
The CAR agonist TCPOBOP inhibits lipogenesis and promotes fibrosis in the mammary gland of adolescent female mice.
Toxicology letters (2018-03-20)
Pengfei Xu, Fan Hong, Jing Wang, Shu Dai, Jialin Wang, Yonggong Zhai
ABSTRAKT
Constitutive androstane receptor (CAR) is a nuclear receptor that not only regulates drug-metabolizing enzymes but also influences energy metabolism. TC, 1, 4-bis [2-(3, 5-dichloropyridyloxy)] benzene (TCPOBOP) has been shown to inhibit lipogenesis in the liver and adipose tissues. The mammary gland is mainly composed of fat pads and duct systems in adolescent female mice. Here, activation of CAR by TC reduces the mammary gland weight, blocks lipid accumulation by inhibiting lipogenesis and gluconeogenesis, and accelerates collagen formation and fibrosis in the mammary fat pad of adolescent female mice. This information provides a reference for CAR activation, which may affect mammary gland development in adolescent females.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TCPOBOP, ≥98% (HPLC), solid
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O