Merck
MARCKSL1 Regulates Spine Formation in the Amygdala and Controls the Hypothalamic-Pituitary-Adrenal Axis and Anxiety-Like Behaviors.
EBioMedicine (2018-03-28)
Takashi Tanaka, Shoko Shimizu, Masaki Ueno, Yoshitaka Fujihara, Masahito Ikawa, Shingo Miyata
ABSTRAKT
Abnormalities in limbic neural circuits have been implicated in the onset of anxiety disorders. However, the molecular pathogenesis underlying anxiety disorders remains poorly elucidated. Here, we demonstrate that myristoylated alanine-rich C-kinase substrate like 1 (MARCKSL1) regulates amygdala circuitry to control the activity of the hypothalamic-pituitary-adrenal (HPA) axis, as well as induces anxiety-like behaviors in mice. MARCKSL1 expression was predominantly localized in the prefrontal cortex (PFC), hypothalamus, hippocampus, and amygdala of the adult mouse brain. MARCKSL1 transgenic (Tg) mice exhibited anxiety-like behaviors dependent on corticotropin-releasing hormone. MARCKSL1 increased spine formation in the central amygdala, and downregulation of MARCKSL1 in the amygdala normalized both increased HPA axis activity and elevated anxiety-like behaviors in Tg mice. Furthermore, MARCKSL1 expression was increased in the PFC and amygdala in a brain injury model associated with anxiety-like behaviors. Our findings suggest that MARCKSL1 expression in the amygdala plays an important role in anxiety-like behaviors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human MARCKSL1