Merck
Cell-extrinsic hematopoietic impact of Ezh2 inactivation in fetal liver endothelial cells.
Blood (2018-03-21)
Wen Hao Neo, Christopher A G Booth, Emanuele Azzoni, Lijun Chi, Paul Delgado-Olguín, Marella F T R de Bruijn, Sten Eirik W Jacobsen, Adam J Mead
ABSTRAKT
Despite the well-established cell-intrinsic role of epigenetic factors in normal and malignant hematopoiesis, their cell-extrinsic role remains largely unexplored. Herein we investigated the hematopoietic impact of inactivating Ezh2, a key component of polycomb repressive complex 2 (PRC2), in the fetal liver (FL) vascular niche. Hematopoietic specific (Vav-iCre) Ezh2 inactivation enhanced FL hematopoietic stem cell (HSC) expansion with normal FL erythropoiesis. In contrast, endothelium (Tie2-Cre) targeted Ezh2 inactivation resulted in embryonic lethality with severe anemia at embryonic day 13.5 despite normal emergence of functional HSCs. Ezh2-deficient FL endothelium overexpressed Mmp9, which cell-extrinsically depleted the membrane-bound form of Kit ligand (mKitL), an essential hematopoietic cytokine, in FL. Furthermore, Mmp9 inhibition in vitro restored mKitL expression along with the erythropoiesis supporting capacity of FL endothelial cells. These data establish that Ezh2 is intrinsically dispensable for FL HSCs and provides proof of principle that modulation of epigenetic regulators in niche components can exert a marked cell-extrinsic impact.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Decamethylcyclopentasiloxane, 97%
Sigma-Aldrich
2,7-Diaminofluorene, >97%
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)