Merck
  • Urethane-based low-temperature curing, highly-customized and multifunctional poly(glycerol sebacate)-co-poly(ethylene glycol) copolymers.
Urethane-based low-temperature curing, highly-customized and multifunctional poly(glycerol sebacate)-co-poly(ethylene glycol) copolymers.
Acta biomaterialia (2018-03-20)
Zihao Wang, Yifan Ma, YanXiang Wang, Yutong Liu, Kai Chen, Zihan Wu, Shuang Yu, Yuan Yuan, Changsheng Liu
ABSTRAKT
Poly (glycerol sebacate) (PGS), a tough elastomer, has been widely explored in tissue engineering due to the desirable mechanical properties and biocompatibility. However, the complex curing procedure (high temperature and vacuum) and limited hydrophilicity (∼90° of wetting angle) greatly impede its functionalities. To address these challenges, a urethane-based low-temperature setting, PEGylated PGS bioelastomer was developed with and without solvent. By simultaneously tailoring PEG and hexamethylene diisocyanate (HDI) contents, the elastomers X-P-mUs (X referred to the PEG content and m referred to HDI content) with a broad ranging mechanical properties and customized hydrophilicity were constructed. The X-P-mUs synthesized exhibited adjustable tensile Young's modulus, ultimate tensile strength and elongation at break in the range of 1.0 MPa-14.2 MPa, 0.3 MPa-7.6 MPa and 53.6%-272.8%, with the water contact angle varying from 28.6° to 71.5°, respectively. Accordingly, these elastomers showed favorable biocompatibility in vitro and mild host response in vivo. Furthermore, the potential applications of X-P-mU elastomers prepared with solvent-base and solvent-free techniques in biomedical fields were investigated. The results showed that these X-P-mU elastomers with high molding capacity at mild temperature could be easily fabricated into various shapes, used as reinforcement for fragile materials, and controllable delivery of drugs and proteins with excellent bioactivity, demonstrating that the X-P-mU elastomers could be tailored as potential building blocks for diverse applications in biomedical research. Poly(glycerol sebacate) (PGS), a tough biodegradable elastomer, has received great attentions in biomedical field. But the complex curing procedure and limited hydrophilicity greatly hamper its functionality. Herein, a urethane-based low-temperature setting, PEGylated PGS (PEGS-U) bioelastomer with highly-customized mechanical properties, hydrophilicity and biodegradability was first explored. The synthesized PEGS-U showed favorable biocompatibility both in vitro and in vivo. Furthermore, the PEGS-U elastomer could be easily fabricated into various shapes, used as reinforcement for fragile materials, and controllable delivery of drugs and proteins with excellent bioactivity. This versatile, user-tunable bioelastomers should be a promising biomaterials for biomedical applications.