Merck
Gold and Female Reproductive Organs: an Ultrastructural Study.
Biological trace element research (2017-08-26)
Nedra Badri, Maroua Mhamdi, Ridha Ben Ali, Horea Matei, Walid-Habib Tekaya, Adrian Florea, Samira Maghraoui, Leila Tekaya
ABSTRAKT
Gold, a heavy yellow-colored metal, is usually found in nature as a metallic element or as salts. This noble metal historically had a reputation as an anti-inflammatory medicine for rheumatoid arthritis, a nervine, and a remedy for nervous disorders, as well as a potential anticancer agent. It has also been used as component in dental restorations and in implant materials. The present study was undertaken to point out histological and ultrastructural effects of gold, administered by intraperitoneal route, in pregnant female reproductive organs (ovary and uterus), in order to clarify its side effects on the reproductive function. Using the transmission electron microscopy (TEM), the ultrastructural investigations of both ultrathin ovarian and uterine sections of treated pregnant rats revealed the existence of numerous heterogeneous clusters with very electron-dense inclusions characterized by various aspects in the lysosomes of granulosa, theca interna cells, and theca externa cells. Degeneration of these tissues, like cell vacuolization, marked expansion of the endoplasmic reticulum, mitochondrial alterations, and necrotic foci, were also highlighted. Moreover, huge phagolysosomes and high numbers of eosinophils as signs of inflammation were also identified especially in endometrial and myometrial cells of gold-treated rats. The ultrastructural investigations of reproductive organ sections of control pregnant rats showed a normal ultrastructural aspect and no loaded lysosomes. These results speculated the toxicity of gold at the used dose. The observed signs of toxicity allowed concluding that the important role of lysosome in the sequestration of this element under an insoluble form in all categories of cells in the studied tissues does not seem to be efficient.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D.E.R. 332, used as embedding medium