Merck
GRO: a novel chemotactic cytokine.
Advances in experimental medicine and biology (1991-01-01)
R Sager, S Haskill, A Anisowicz, D Trask, M C Pike
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
KC (CXCL1) from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture