Merck
Molecular cloning and expression of biologically active human glia maturation factor-beta.
Journal of neurochemistry (1991-08-01)
R Kaplan, A Zaheer, M Jaye, R Lim
ABSTRAKT
Glia maturation factor-beta, a protein found in the brains of all vertebrates thus far examined, appears to play a role in the differentiation, maintenance, and regeneration of the nervous system. Using oligonucleotide probes based on the sequences of three tryptic peptides derived from bovine glia maturation factor-beta, we screened a human brainstem cDNA library in lambda gt11. A 0.7-kb clone was isolated, sequenced in its entirety, and found to encode a polypeptide of 142 amino acids which contained regions identical to the three bovine peptides. This polypeptide, human recombinant glia maturation factor-beta, has been expressed in Escherichia coli and found to possess structural characteristics and biological activity indistinguishable from those of the native bovine protein.