Merck
  • Biologically active polymers: synthesis and antimicrobial activity of modified glycidyl methacrylate polymers having a quaternary ammonium and phosphonium groups.
Biologically active polymers: synthesis and antimicrobial activity of modified glycidyl methacrylate polymers having a quaternary ammonium and phosphonium groups.
Journal of Controlled Release : Official Journal of the Controlled Release Society (1998)
Kenawy E-R, et al.