Merck
  • One-step synthesis of copper nanoparticles containing mesoporous silica by nanocasting of binuclear copper (II) complexes with cyclodextrins.
One-step synthesis of copper nanoparticles containing mesoporous silica by nanocasting of binuclear copper (II) complexes with cyclodextrins.
Journal of Materials Chemistry (2003)
Han B H and Antonietti M