Merck
Granzymes at a glance.
Journal of cell science (2006-12-13)
Michael Bots, Jan Paul Medema
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Granzyme B from mouse, recombinant, expressed in Hi-5 Insect cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture