Merck
Mithramycin A Alleviates Osteoarthritic Cartilage Destruction by Inhibiting HIF-2α Expression.
International journal of molecular sciences (2018-05-12)
Moon-Chang Choi, Woo Hee Choi
ABSTRAKT
Osteoarthritis (OA) is the most common and increasing joint disease worldwide. Current treatment for OA is limited to control of symptoms. The purpose of this study was to determine the effect of specificity protein 1 (SP1) inhibitor Mithramycin A (MitA) on chondrocyte catabolism and OA pathogenesis and to explore the underlying molecular mechanisms involving SP1 and other key factors that are critical for OA. Here, we show that MitA markedly inhibited expressions of matrix-degrading enzymes induced by pro-inflammatory cytokine interleukin-1β (IL-1β) in mouse primary chondrocytes. Intra-articular injection of MitA into mouse knee joint alleviated OA cartilage destruction induced by surgical destabilization of the medial meniscus (DMM). However, modulation of SP1 level in chondrocyte and mouse cartilage did not alter catabolic gene expression or cartilage integrity, respectively. Instead, MitA significantly impaired the expression of HIF-2α known to be critical for OA pathogenesis. Such reduction in expression of HIF-2α by MitA was caused by inhibition of NF-κB activation, at least in part. These results suggest that MitA can alleviate OA pathogenesis by suppressing NF-κB-HIF-2α pathway, thus providing insight into therapeutic strategy for OA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human SP1