Merck
  • Differential effect of activin on mouse embryonic stem cell differentiation in insulin?secreting cells under nestin?positive selection and spontaneous differentiation protocols.
Differential effect of activin on mouse embryonic stem cell differentiation in insulin?secreting cells under nestin?positive selection and spontaneous differentiation protocols.
Cell Biology International (2008)
Jafary H, et al.