Merck
Three new abietane-type diterpenoids from the leaves of Indonesian Plectranthus scutellarioides.
Fitoterapia (2018-02-16)
Takuya Ito, Senda Kartika Rakainsa, Khoirun Nisa, Hiroyuki Morita
ABSTRAKT
Three new diterpenoids, spiroscutelones A-C (1-3), along with known diterpene 4, were isolated from the leaves of Plectranthus scutellarioides. Their structures were established based on extensive spectroscopic analyses, including MS and NMR spectroscopy. Spiroscutelone A (1) represents the first example of an abietane-type terpenoid skeleton with a cyclobutane moiety linking the B and C rings. Of compounds 1-4, spiroscutelone B (2) exhibited the most potent cytotoxicities against the three tested human cancer lines [breast (MCF-7), pancreatic (PSN-1), and cervical (HeLa)] with IC50 values ranging from 17.9 μM to 29.8 μM and low cytotoxicity against a lung fibroblast (WI-38) normal cell line.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%