Merck
PINK1/Parkin-mediated mitophagy was activated against 1,4-Benzoquinone-induced apoptosis in HL-60 cells.
Toxicology in vitro : an international journal published in association with BIBRA (2018-03-24)
Chunxiao Zhang, Xiuyuan Yu, Jiahao Gao, Qianqian Zhang, Shuqiang Sun, Hua Zhu, Xinjun Yang, Hongtao Yan
ABSTRAKT
Hematotoxicity of benzene is derived mainly from its active metabolite, 1,4-Benzoquinone (1,4-BQ), which induces cell apoptosis and mitochondrial damage. Damaged mitochondria are degraded through a specialized autophagy pathway, called mitophagy, which is driven by PINK1/Parkin signaling. However, whether mitophagy is involved in 1,4-BQ-induced toxicity remains unclear. This study was designed to investigate whether PINK1/Parkin-mediated mitophagy is activated in 1,4-BQ-treated HL-60 cells, and the roles mitophagy plays in 1,4-BQ-induced apoptosis. Our results demonstrated that 1,4-BQ induced autophagy in HL-60 cells, characterized by increased LC3-II/LC3-I ratio and Beclin1 expression, as well as decreased expression of p62. We confirmed the presence of mitophagosomes using electron microscopy, and found that 1,4-BQ-induced autophagy was blocked by pretreatment with the mitophagy inhibitor Cyclosporine A (CsA). In addition, we found that 1,4-BQ induced mitochondrial stress through decreased mitochondrial membrane potential (MMP) and increasedproduction of reactive oxygen species (ROS). We also confirmed that 1,4-BQ-induced mitophagy was mediated by the PINK1/Parkin pathway, illustrated by increased expression of PINK1 and Parkin mRNA and protein. Finally, we examined 1,4-BQ-induced apoptosis with or without CsA, which demonstrated that apoptosis increased after mitophagy inhibition, suggesting that mitophagy has a protective effect in this context. In conclusion, this study demonstrates that the activated PINK1/Parkin-mediated mitophagy exerts a significantly protective effect against 1,4-BQ-induced apoptosis in HL-60 cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
JC-1, powder or solid (Crystals)