Merck
  • Total resection of any segment of the lateral meniscus may cause early cartilage degeneration: Evaluation by magnetic resonance imaging using T2 mapping.
Total resection of any segment of the lateral meniscus may cause early cartilage degeneration: Evaluation by magnetic resonance imaging using T2 mapping.
Medicine (2018-06-08)
Koji Murakami, Yuji Arai, Kazuya Ikoma, Kammei Kato, Hiroaki Inoue, Shuji Nakagawa, Yuta Fujii, Keiichiro Ueshima, Hiroyoshi Fujiwara, Toshikazu Kubo
ABSTRAKT
The aim of this study was to perform quantitative evaluation of degeneration of joint cartilage using T2 mapping in magnetic resonance imaging (MRI) after arthroscopic partial resection of the lateral meniscus.The subjects were 21 patients (23 knees) treated with arthroscopic partial resection of the lateral meniscus. MRI was performed for all knees before surgery and 6 months after surgery to evaluate the center of the lateral condyle of the femur in sagittal images for T2 mapping. Ten regions of interest (ROIs) on the articular cartilage were established at 10-degree intervals, from the point at which the femur shaft crossed the lateral femoral condyle joint to the articular cartilage 90° relative to the femur shaft. Preoperative and postoperative T2 values were evaluated at each ROI. Age, sex, body mass index, femorotibial angle, Tegner score, and amount of meniscal resection were evaluated when the T2 value increased more than 6% at 30°.T2 values at approximately 10 °, 20 °, 30 °, 40 °, 50 °, and 60 ° degrees relative to the anatomical axis of the femur were significantly greater postoperatively (3.1, 3.6, 5.5, 4.4, 5.0, 6.4%, respectively) than preoperatively. A >6% increase at 30° was associated with total resection of any segment of the meniscus.Degeneration of the articular cartilage, as shown by the disorganization of collagen arrays at positions approximately 10 °, 20 °, 30 °, 40 °, 50 °, and 60 ° relative to the anatomical axis of the femur, may start soon after arthroscopic lateral meniscectomy. Total resection of any segment of the lateral meniscus may cause T2 elevation of articular cartilage of lateral femoral condyle.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)