Merck
Loss of expression and prognosis value of alpha-internexin in gastroenteropancreatic neuroendocrine neoplasm.
BMC cancer (2018-06-27)
Yuhong Wang, Yuanjia Chen, Xiaoxing Li, Wanming Hu, Yu Zhang, Luohai Chen, Minhu Chen, Jie Chen
ABSTRAKT
The neuronal intermediate filament alpha-internexin (α-internexin) is a cytoskeleton protein which is involved in the tumor initiation and progression. In this study, we examined the expression and prognosis value of α-internexin in gastroenteropancreatic neuroendocrine neoplasms (GEP-NENs). α-internexin was detected with immunohistochemical staining in 286 tumor specimens from patients with GEP-NENs. Methylation status of α-internexin was evaluated by bisulfite genomic sequencing. We assessed the prognostic value of α-internexin and its correlation with relevant clinicalpathological characteristics. The reduced/loss of expression rate of α-internexin in GEP-NEN was 73.4% (210/286), while the positive expression rate was 26.6% (76/286). The difference of α-internexin deficiency was not statistically significant between gastrointestinal NENs (GI-NENs) and pancreatic NENs (pNENs). However, we found significant difference of reduced/loss of α-internexin expression among different sites of GI-NENs (χ2 = 43.470, P < 0.001). The reduced/loss of expression of α-internexin was significantly associated with poorly differentiation (P < 0.001) and advanced tumor stage (P < 0.001). Univariate analyses showed that reduced/loss of expression of α-internexin predicted worse overall survival (OS) in GEP-NEN patients (P < 0.001), especially in subtype of GI-NENs (P < 0.001). However, in multivariable regression analysis, α-internexin expression was not an independent prognostic factor. The hypermethylation of α-internexin gene was significantly correlated with protein deficiency in GI-NENs, but not in pNENs. Hypermethylation of several CpG sites was significantly associated with poorly differentiated and advanced stage (P values range from 0.018 to 0.044). However, the methylation status of α-internexin was not associated with patient OS. The expression of α-internexin was highly heterougeneous in different sites of GEP-NENs. The reduced/loss of expression of α-internexin was closely related to tumors with aggressiveness and patient's adverse prognosis. The hypermethylation of the regulatory region examined may be an important epigenetic regulation mechanism of α-internexin deficiency in subtype of GI-NENs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%