Merck
Highly efficient G-quadruplex recognition by bisquinolinium compounds.
Journal of the American Chemical Society (2007-01-31)
Anne De Cian, Elsa Delemos, Jean-Louis Mergny, Marie-Paule Teulade-Fichou, David Monchaud
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PhenDC3, ≥97% (HPLC)