Merck
  • Optimization of 6,6-dimethyl pyrrolo[3,4-c]pyrazoles: Identification of PHA-793887, a potent CDK inhibitor suitable for intravenous dosing.
Optimization of 6,6-dimethyl pyrrolo[3,4-c]pyrazoles: Identification of PHA-793887, a potent CDK inhibitor suitable for intravenous dosing.
Bioorganic & medicinal chemistry (2010-02-16)
Maria Gabriella Brasca, Clara Albanese, Rachele Alzani, Raffaella Amici, Nilla Avanzi, Dario Ballinari, James Bischoff, Daniela Borghi, Elena Casale, Valter Croci, Francesco Fiorentini, Antonella Isacchi, Ciro Mercurio, Marcella Nesi, Paolo Orsini, Wilma Pastori, Enrico Pesenti, Paolo Pevarello, Patrick Roussel, Mario Varasi, Daniele Volpi, Anna Vulpetti, Marina Ciomei
ABSTRAKT
We have recently reported CDK inhibitors based on the 6-substituted pyrrolo[3,4-c]pyrazole core structure. Improvement of inhibitory potency against multiple CDKs, antiproliferative activity against cancer cell lines and optimization of the physico-chemical properties led to the identification of highly potent compounds. Compound 31 (PHA-793887) showed good efficacy in the human ovarian A2780, colon HCT-116 and pancreatic BX-PC3 carcinoma xenograft models and was well tolerated upon daily treatments by iv administration. It was identified as a drug candidate for clinical evaluation in patients with solid tumors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PHA-793887, ≥98% (HPLC)