Merck
  • Preparation of thiol- and amine-bifunctionalized hybrid monolithic column via "one-pot" and applications in speciation of inorganic arsenic.
Preparation of thiol- and amine-bifunctionalized hybrid monolithic column via "one-pot" and applications in speciation of inorganic arsenic.
Talanta (2018-10-24)
Ling-Yu Zhao, Qing-Yun Zhu, Li Mao, Yi-Jun Chen, Hong-Zhen Lian, Xin Hu
ABSTRAKT
A thiol- and amine-bifunctionalized organic-inorganic hybrid monolithic column was prepared for the first time via one-pot co-condensation of 3-mercaptopropyltrimethoxysilane, N-(2-aminoethyl)-3-aminopropyltriethoxysilane and tetraethylorthosilicate, and utilized for separation and enrichment of inorganic arsenic species. Various parameters of solid phase microextraction (SPME) operation and analytical performance were also investigated systematically. Under the optimum condition, both As(III) and As(V) can be adsorbed over a wide pH range (3.0-8.0) and eluted in turn, in which 3% HNO3 (v/v) was firstly used to selectively release As(V), and then 3% HNO3 with 0.01 mol L-1 KIO3 or 15% mercaptosuccinic acid (MSA) (m/v) to selectively release As(III). Meanwhile, the elution mechanism of As(III) and As(V) was elucidated comprehensively, and notably, the novel eluent, HNO3 + MSA, was recommended for eluting As(III). Therefore, the thiol- and amine-bifunctionalized organic-inorganic hybrid monolithic column as an ideal SPME matrix for the speciation analysis of arsenic in environmental waters has the merits of facile preparation, low cost, high adsorption capacity and selective desorption. In addition, compared with thiol- and amine-bifunctionalized mesoporous silica, the bifunctional hybrid monolith based SPME protocol with less time and reagent consumption is promising to be applied not only to filed sampling but also to on-line analysis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub