Merck
The nonreceptor tyrosine kinase c-Abl phosphorylates Runx1 and regulates Runx1-mediated megakaryocyte maturation.
Biochimica et biophysica acta. Molecular cell research (2018-05-08)
Hainan Liu, Yan Cui, Guang-Fei Wang, Qincai Dong, Yebao Yao, Ping Li, Cheng Cao, Xuan Liu
ABSTRAKT
The transcription factor Runx1 is an essential regulator of definitive hematopoiesis, megakaryocyte (MK) maturation, and lymphocyte differentiation. Runx1 mutations that interfere with its transcriptional activity are often present in leukemia patients. Recent work demonstrated that the transcriptional activity of Runx1 is regulated by kinase-mediated phosphorylation. In this study, we showed that c-Abl, but not Arg tyrosine kinase, associated with Runx1 both in cultured cells and in vitro. c-Abl-mediated tyrosine phosphorylation in the Runx1 transcription inhibition domain negatively regulated the transcriptional activity of Runx1 and inhibited Runx1-mediated MK maturation. Consistent with these findings, increased numbers of MKs were detected in the spleens and bone marrow of abl gene conditional knockout mice. Our findings demonstrate an important role of c-Abl kinase in Runx1-mediated MK maturation and platelet formation and provide a potential mechanism of Abl kinase-regulated hematopoiesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection