Merck
Activation of beta-catenin-Tcf signaling in colon cancer by mutations in beta-catenin or APC.
Science (New York, N.Y.) (1997-03-21)
P J Morin, A B Sparks, V Korinek, N Barker, H Clevers, B Vogelstein, K W Kinzler
ABSTRAKT
Inactivation of the adenomatous polyposis coli (APC) tumor suppressor gene initiates colorectal neoplasia. One of the biochemical activities associated with the APC protein is down-regulation of transcriptional activation mediated by beta-catenin and T cell transcription factor 4 (Tcf-4). The protein products of mutant APC genes present in colorectal tumors were found to be defective in this activity. Furthermore, colorectal tumors with intact APC genes were found to contain activating mutations of beta-catenin that altered functionally significant phosphorylation sites. These results indicate that regulation of beta-catenin is critical to APC's tumor suppressive effect and that this regulation can be circumvented by mutations in either APC or beta-catenin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Catenin β, GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution