Merck
  • Dissociable effects of mGluR5 allosteric modulation on distinct forms of impulsivity in rats: interaction with NMDA receptor antagonism.
Dissociable effects of mGluR5 allosteric modulation on distinct forms of impulsivity in rats: interaction with NMDA receptor antagonism.
Psychopharmacology (2015-06-13)
Sarah N Isherwood, Anton Pekcec, Janet R Nicholson, Trevor W Robbins, Jeffrey W Dalley
ABSTRAKT
Impaired N-methyl-D-aspartate (NMDA) receptor signalling underlies several psychiatric disorders that express high levels of impulsivity. Although synergistic interactions exist between NMDA receptors and metabotropic glutamate receptor 5 (mGluR5), the significance of this interaction for impulsivity is unknown. This study aims to investigate the effects of negative and positive allosteric mGluR5 modulation (NAM/PAM) on trait impulsivity and impulsivity evoked by NMDA receptor antagonism in rats. Motor and choice impulsivity were assessed using the five-choice serial reaction time task (5-CSRTT) and delayed-discounting task (DDT), respectively. The effects of RO4917523 and 3-[(2-methyl-1,3-thiazol-4-yl)ethynyl]pyridine (MTEP) (NAMs) and ADX47273 (PAM) were investigated in non-impulsive rats and in trait high- and low-impulsive rats. The effects of these compounds on impulsivity induced by NMDA receptor antagonism (MK801) in the 5-CSRTT were also investigated. RO4917523 (0.1-1 mg/kg) decreased premature responding and increased omissions but had no effect on locomotor activity up to 0.1 mg/kg. MTEP significantly increased omissions, decreased accuracy and slowed responding but had no effect on premature responding. ADX47273 decreased premature responding at doses that had no effect on locomotor activity. MK801 increased premature responding and impaired attentional accuracy; these deficits were dose dependently rescued by ADX47273 pre-treatment. Allosteric modulation of mGluR5 had no significant effect on choice impulsivity, nor did it modulate general task performance. These findings demonstrate that mGluR5 allosteric modulation selectively dissociates motor and choice impulsivity. We further show that mGluR5 PAMs may have therapeutic utility in selectively targeting specific aspects of impulsivity and executive dysfunction.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ADX-47273, ≥98% (HPLC)