Merck
Histone H3 Thr-3 phosphorylation by Haspin positions Aurora B at centromeres in mitosis.
Science (New York, N.Y.) (2010-08-14)
Fangwei Wang, Jun Dai, John R Daum, Ewa Niedzialkowska, Budhaditya Banerjee, P Todd Stukenberg, Gary J Gorbsky, Jonathan M G Higgins
ABSTRAKT
Aurora B is a component of the chromosomal passenger complex (CPC) required for correct spindle-kinetochore attachments during chromosome segregation and for cytokinesis. The chromatin factors that recruit the CPC to centromeres are unknown, however. Here we show that phosphorylation of histone H3 threonine 3 (H3T3ph) by Haspin is necessary for CPC accumulation at centromeres and that the CPC subunit Survivin binds directly to H3T3ph. A nonbinding Survivin-D70A/D71A mutant does not support centromeric CPC concentration, and both Haspin depletion and Survivin-D70A/D71A mutation diminish centromere localization of the kinesin MCAK and the mitotic checkpoint response to taxol. Survivin-D70A/D71A mutation and microinjection of H3T3ph-specific antibody both compromise centromeric Aurora B functions but do not prevent cytokinesis. Therefore, H3T3ph generated by Haspin positions the CPC at centromeres to regulate selected targets of Aurora B during mitosis.