Merck
  • The Streptomyces lividans family 12 endoglucanase: construction of the catalytic core, expression, and X-ray structure at 1.75 ? resolution.
The Streptomyces lividans family 12 endoglucanase: construction of the catalytic core, expression, and X-ray structure at 1.75 ? resolution.
Biochemistry (1997)
Sulzenbacher G, et al.