Merck
Phosphatidylinositol 4 phosphate regulates targeting of clathrin adaptor AP-1 complexes to the Golgi.
Cell (2003-08-14)
Ying Jie Wang, Jing Wang, Hui Qiao Sun, Manuel Martinez, Yu Xiao Sun, Eric Macia, Tomas Kirchhausen, Joseph P Albanesi, Michael G Roth, Helen L Yin
ABSTRAKT
Phosphatidylinositol 4 phosphate [PI(4)P] is essential for secretion in yeast, but its role in mammalian cells is unclear. Current paradigms propose that PI(4)P acts primarily as a precursor to phosphatidylinositol 4,5 bisphosphate (PIP2), an important plasma membrane regulator. We found that PI(4)P is enriched in the mammalian Golgi, and used RNA interference (RNAi) of PI4KIIalpha, a Golgi resident phosphatidylinositol 4 kinase, to determine whether PI(4)P directly regulates the Golgi. PI4KIIalpha RNAi decreases Golgi PI(4)P, blocks the recruitment of clathrin adaptor AP-1 complexes to the Golgi, and inhibits AP-1-dependent functions. This AP-1 binding defect is rescued by adding back PI(4)P. In addition, purified AP-1 binds PI(4)P, and anti-PI(4)P inhibits the in vitro recruitment of cytosolic AP-1 to normal cellular membranes. We propose that PI4KIIalpha establishes the Golgi's unique lipid-defined organelle identity by generating PI(4)P-rich domains that specify the docking of the AP-1 coat machinery.