Merck
Preventive treatment with liraglutide protects against development of glucose intolerance in a rat model of Wolfram syndrome.
Scientific reports (2018-07-07)
Maarja Toots, Kadri Seppa, Toomas Jagomäe, Tuuliki Koppel, Maia Pallase, Indrek Heinla, Anton Terasmaa, Mario Plaas, Eero Vasar
ABSTRAKT
Wolfram syndrome (WS) is a rare autosomal recessive disorder caused by mutations in the WFS1 (Wolframin1) gene. The syndrome first manifests as diabetes mellitus, followed by optic nerve atrophy, deafness, and neurodegeneration. The underlying mechanism is believed to be a dysregulation of endoplasmic reticulum (ER) stress response, which ultimately leads to cellular death. Treatment with glucagon-like peptide-1 (GLP-1) receptor agonists has been shown to normalize ER stress response in several in vitro and in vivo models. Early chronic intervention with the GLP-1 receptor agonist liraglutide starting before the onset of metabolic symptoms prevented the development of glucose intolerance, improved insulin and glucagon secretion control, reduced ER stress and inflammation in Langerhans islets in Wfs1 mutant rats. Thus, treatment with GLP-1 receptor agonists might be a promising strategy as a preventive treatment for human WS patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Liraglutide, ≥95% (HPLC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid