Merck
Mycophenolate mofetil sensor based on molecularly imprinted polymer/multi-walled carbon nanotubes modified carbon paste electrode.
Analytical biochemistry (2018-07-22)
Hossein Momeneh, Mohammad Bagher Gholivand
ABSTRAKT
Using square wave voltammetry, a carbon paste electrode modified by molecularly imprinted polymer (MIP) as a recognition element of mycophenolate mofetil (MMF) and multi-walled carbon nanotubes was used for MMF monitoring To investigate the electrode during modification, electrochemical impedance spectroscopy (EIS) and cyclic voltammetry (CV) were utilized. After optimization of the effective parameters, the anodic peak current of MMF was utilized for dynamic range study which was linear in 9.9 nM-87 μM range. The detection limit of the sensor was 7.0 nM. The capture ability of MIP to target was compared with that of non-imprinted polymer (NIP). The practical application of the sensor in biological fluid samples analysis demonstrates its selectivity, sensitivity, and stability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor