Merck
  • Generation of a human induced pluripotent stem cell line (LL008 1.4) from a familial Alzheimer's disease patient carrying a double KM670/671NL (Swedish) mutation in APP gene.
Generation of a human induced pluripotent stem cell line (LL008 1.4) from a familial Alzheimer's disease patient carrying a double KM670/671NL (Swedish) mutation in APP gene.
Stem cell research (2018-08-14)
Minna Oksanen, Ida Hyötyläinen, Jenni Voutilainen, Katja A Puttonen, Riikka H Hämäläinen, Caroline Graff, Šárka Lehtonen, Jari Koistinaho
ABSTRAKT
A double mutation (KM670/671NL) in amyloid precursor protein gene (APP) is causative for familial Alzheimer's disease and has been shown to increase the total Aβ burden. Here we report the generation and characterization of an iPSC line from a fAD patient carrying APP KM670/671NL. The generated iPSCs retained the mutation, expressed pluripotency markers, showed a normal karyotype and differentiated into all three germ layers. This iPSC line can be used, for example, in disease modeling and mechanistic studies. Resource table.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-α-Fetoprotein (AFP) antibody produced in mouse, ascites fluid, clone C3
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, clone TRA-1-81, Chemicon®, from mouse