Merck
  • Effect of Sodium Selenate on Hippocampal Proteome of 3×Tg-AD Mice-Exploring the Antioxidant Dogma of Selenium against Alzheimer's Disease.
Effect of Sodium Selenate on Hippocampal Proteome of 3×Tg-AD Mice-Exploring the Antioxidant Dogma of Selenium against Alzheimer's Disease.
ACS chemical neuroscience (2018-04-12)
Javed Iqbal, Kaoyuan Zhang, Na Jin, Yuxi Zhao, Qiong Liu, Jiazuan Ni, Liming Shen
ABSTRAKT
Selenium (Se), an antioxidant trace element, is an important nutrient for maintaining brain functions and is reported to be involved in Alzheimer's disease (AD) pathologies. The present study has been designed to elucidate the protein changes in hippocampus of 3×Tg-AD mice after supplementing sodium selenate as an inorganic source of selenium. By using iTRAQ proteomics technology, 113 differentially expressed proteins (DEPs) are found in AD/WT mice with 37 upregulated and 76 downregulated proteins. Similarly, in selenate-treated 3×Tg-AD (ADSe/AD) mice, 115 DEPs are found with 98 upregulated and 17 downregulated proteins. The third group of mice (ADSe/WT) showed 75 DEPs with 46 upregulated and 29 downregulated proteins. Among these results, 42 proteins (40 downregulated and 2 upregulated) in the diseased group showed reverse expression when treated with selenate. These DEPs are analyzed with different bioinformatics tools and are found associated with various AD pathologies and pathways. Based on their functions, selenate-reversed proteins are classified as structural proteins, metabolic proteins, calcium regulating proteins, synaptic proteins, signaling proteins, stress related proteins, and transport proteins. Six altered AD associated proteins are successfully validated by Western blot analysis. This study shows that sodium selenate has a profound effect on the hippocampus of the triple transgenic AD mice. This might be established as an effective therapeutic agent after further investigation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)