Merck
Chemokine CC-motif ligand 2 participates in platelet function and arterial thrombosis by regulating PKCα-P38MAPK-HSP27 pathway.
Biochimica et biophysica acta. Molecular basis of disease (2018-06-05)
Dan Liu, Yu Cao, Xiaolin Zhang, Chengfei Peng, Xiaoxiang Tian, Chenghui Yan, Yanxia Liu, Meili Liu, Yaling Han
ABSTRAKT
Studies indicate that chemokine CC-motif ligand 2 (CCL2) is involved in inflammation and atherosclerosis. However, the roles and mechanisms of CCL2 on platelet function and arterial thrombosis are unknown. The expressions of CCL2 or CCR2 in the plasma, platelets and coronary thrombus of ST-elevated myocardial infarction (STEMI) patients were examined by ELISA, Western blot, immunohistochemistry and immunofluorescence. The roles of CCL2 on platelet aggregation, activation and secretion were examined by light transmission aggregometry, flow cytometry and ELISA. The expressions of CCL2 or CCR2 in the plasma or platelets of STEMI patients with platelet high response were higher than those with platelet normal response; In vitro, exogenous recombinant human CCL2 markedly increased platelet aggregation, activation and granule secretion, which were abolished by CCL2 neutralizing antibody or CCR2 inhibiter. CCL2 increased the phosphorylation levels of PKCα (Thr638), P38MAPK (Thr180/Tyr182) and HSP27 (S78/S82) in human platelets, which were abrogated by PKCα inhibitor (RO 318220) or P38MAPK inhibitor (SB 203580). RO 318220 or SB 203580 diminished CCL2-induced platelet function. In CCL2-/- mice, platelet aggregation and secretion were attenuated; the phosphorylation of PKCα, P38MAPK and HSP27 were decreased. In a carotid arterial thrombus mouse model, CCL2-/- mice displayed a significantly extended carotid artery occlusion time compared with wild type. CCL2 played important roles in regulating platelet function and arterial thrombosis through the PKCα-P38MAPK-HSP27 pathway, which might provide theoretical basis for searching new antiplatelet drugs and the treatment for cardiovascular diseases.