Merck
VprBP (DCAF1) Regulates RAG1 Expression Independently of Dicer by Mediating RAG1 Degradation.
Journal of immunology (Baltimore, Md. : 1950) (2018-06-22)
N Max Schabla, Greg A Perry, Victoria L Palmer, Patrick C Swanson
ABSTRAKT
The assembly of Ig genes in developing B lymphocytes by V(D)J recombination is initiated by the RAG1-RAG2 endonuclease complex. We previously identified an interaction between RAG1 and viral protein R binding protein (VprBP) (also known as DNA damage binding protein 1 cullin 4-associated factor 1 [DCAF1]), a substrate receptor for the cullin 4-really interesting new gene (RING) E3 ubiquitin ligase (CRL4). We report in this article that in mice, B cell-intrinsic loss of VprBP increases RAG1 protein levels and disrupts expression of the endoribonuclease Dicer, which is essential for microRNA maturation. Rag1/2 transcription is known to be derepressed by loss of microRNA-mediated suppression of phosphatase and tensin homolog, raising the possibility that the elevated level of RAG1 observed in VprBP-deficient B cells is caused indirectly by the loss of Dicer. However, we show that VprBP restrains RAG1 expression posttranscriptionally and independently of Dicer. Specifically, loss of VprBP stabilizes RAG1 protein, which we show is normally degraded via a mechanism requiring both 20S proteasome and cullin-RING E3 ubiquitin ligase activity. Furthermore, we show that RAG1 stabilization through small molecule inhibition of cullin-RING E3 ubiquitin ligase activation promotes V(D)J recombination in a murine pre-B cell line. Thus, in addition to identifying a role for VprBP in maintaining Dicer levels in B cells, our findings reveal the basis for RAG1 turnover and provide evidence that the CRL4VprBP(DCAF1) complex functions to maintain physiological levels of V(D)J recombination.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Przeciwciało anty-RAG-2, klon 39, culture supernatant, from rabbit