Merck
RNF169 limits 53BP1 deposition at DSBs to stimulate single-strand annealing repair.
Proceedings of the National Academy of Sciences of the United States of America (2018-08-15)
Liwei An, Chao Dong, Junshi Li, Jie Chen, Jingsong Yuan, Jun Huang, Kui Ming Chan, Cheng-Han Yu, Michael S Y Huen
ABSTRAKT
Unrestrained 53BP1 activity at DNA double-strand breaks (DSBs) hampers DNA end resection and upsets DSB repair pathway choice. RNF169 acts as a molecular rheostat to limit 53BP1 deposition at DSBs, but how this fine balance translates to DSB repair control remains undefined. In striking contrast to 53BP1, ChIP analyses of AsiSI-induced DSBs unveiled that RNF169 exhibits robust accumulation at DNA end-proximal regions and preferentially targets resected, RPA-bound DSBs. Accordingly, we found that RNF169 promotes CtIP-dependent DSB resection and favors homology-mediated DSB repair, and further showed that RNF169 dose-dependently stimulates single-strand annealing repair, in part, by alleviating the 53BP1-imposed barrier to DSB end resection. Our results highlight the interplay of RNF169 with 53BP1 in fine-tuning choice of DSB repair pathways.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Mimosine from Koa hoale seeds, ≥98%
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer