Merck
  • The effect of auxin (indole-3-acetic acid) on the growth rate and tropism of the sporangiophore of Phycomyces blakesleeanus and identification of auxin-related genes.
The effect of auxin (indole-3-acetic acid) on the growth rate and tropism of the sporangiophore of Phycomyces blakesleeanus and identification of auxin-related genes.
Protoplasma (2018-03-11)
Branka D Živanović, Kristian K Ullrich, Bianka Steffens, Sladjana Z Spasić, Paul Galland
ABSTRAKT
The roles of fungal auxins in the regulation of elongation growth, photo-, and gravitropism are completely unknown. We analyzed the effects of exogenous IAA (indole-3-acetic acid), various synthetic auxins including 1-NAA (1-naphthaleneacetic acid) and 2,4-D (2,4-dichlorophenoxyacetic acid), and the auxin transport inhibitor NPA (N-1-naphtylphtalamic acid) on the growth rate and bending of the unicellular sporangiophore of the zygomycete fungus, Phycomyces blakesleeanus. Sporangiophores that were submerged in an aqueous buffer responded to IAA with a sustained enhancement of the growth rate, while 1-NAA, 2,4-D, and NPA elicited an inhibition. In contrast, sporangiophores kept in air responded to IAA with a 20 to 40% decrease of the growth rate, while 1-NAA and NPA elicited an enhancement. The unilateral and local application of IAA in the growing zone of the sporangiophore elicited in 30 min a moderate negative tropic bending in wild type C2 and mutant C148madC, which was, however, partially masked by a concomitant avoidance response caused by the aqueous buffer. Auxin transport-related genes ubiquitous in plants were found in a BLAST search of the Phycomyces genome. They included members of the AUX1 (auxin influx carrier protein 1), PILS (PIN-LIKES, auxin transport facilitator protein), and ABCB (plant ATP-binding cassette transporter B) families while members of the PIN family were absent. Our observations imply that IAA represents an intrinsic element of the sensory transduction of Phycomyces and that its mode of action must very likely differ in several respects from that operating in plants.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)