Merck
Supported rhodium oxide nanoparticles as highly active CO oxidation catalysts.
Angewandte Chemie (International ed. in English) (2011-04-22)
D A J Michel Ligthart, Rutger A van Santen, Emiel J M Hensen