Merck
  • GFZF, a glutathione S-transferase protein implicated in cell cycle regulation and hybrid inviability, is a transcriptional co-activator.
GFZF, a glutathione S-transferase protein implicated in cell cycle regulation and hybrid inviability, is a transcriptional co-activator.
Molecular and cellular biology (2017-11-22)
Douglas G Baumann, Mu-Shui Dai, Hua Lu, David S Gilmour
ABSTRAKT
The core promoter of protein-encoding genes plays a central role in regulating transcription. M1BP is a transcriptional activator that associates with a core promoter element known as Motif 1 that resides at thousand of genes in Drosophila. To gain insight into how M1BP functions, we identified an interacting protein called GFZF. GFZF had been previously identified in genetic screens for factors involved in maintenance of hybrid inviability, the G2-M DNA damage checkpoint, and RAS/MAPK signaling but its contribution to these processes was unknown. Here, we show that GFZF resides in the nucleus and functions as a transcriptional co-activator. In addition, we show that GFZF is a glutathione S-transferase(GST). Thus, GFZF is the first transcriptional co-activator with intrinsic GST activity, and its identification as a transcriptional co-activator provides an explanation for its role in numerous biological processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)