Merck
BTBD18 Regulates a Subset of piRNA-Generating Loci through Transcription Elongation in Mice.
Developmental cell (2017-03-16)
Liquan Zhou, Bertram Canagarajah, Yangu Zhao, Boris Baibakov, Keizo Tokuhiro, Dragan Maric, Jurrien Dean
ABSTRAKT
PIWI-interacting RNAs (piRNAs) are small non-coding RNAs essential for animal germ cell development. Despite intense investigation of post-transcriptional processing, chromatin regulators for piRNA biogenesis in mammals remain largely unexplored. Here we document that BTBD18 is a pachytene nuclear protein in mouse testes that occupies a subset of pachytene piRNA-producing loci. Ablation of Btbd18 in mice disrupts piRNA biogenesis, prevents spermiogenesis, and results in male sterility. Transcriptome profiling, chromatin accessibility, and RNA polymerase II occupancy demonstrate that BTBD18 facilitates expression of pachytene piRNA precursors by promoting transcription elongation. Thus, our study identifies BTBD18 as a specific controller for transcription activation through RNA polymerase II elongation at a subset of genomic piRNA loci.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-MYBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H2A.X (Ser139), klon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)