Merck
  • Translocation of forkhead box O1 to the nuclear periphery induces histone modifications that regulate transcriptional repression of PCK1 in HepG2 cells.
Translocation of forkhead box O1 to the nuclear periphery induces histone modifications that regulate transcriptional repression of PCK1 in HepG2 cells.
Genes to cells : devoted to molecular & cellular mechanisms (2015-03-05)
Tamaki Arai, Fumi Kano, Masayuki Murata
ABSTRAKT
Forkhead box O1 (FOXO1) is an important target for insulin. It is widely accepted that insulin-induced phosphorylation of FOXO1 by Akt leads to its nuclear exclusion and results in the inhibition of FOXO1-mediated transcription of the gluconeogenic gene phosphoenolpyruvate carboxykinase 1 (PCK1) in hepatocytes. However, many results that contradict this model have accumulated. Here, we provide a new mechanism for insulin-dependent repression of FOXO1-mediated transcription. We showed insulin-induced translocation of endogenous Ser256-phosphorylated FOXO1, which is essential for regulation of FOXO1-mediated transcription, from nuclear speckles to the nuclear periphery. This insulin-dependent translocation of FOXO1 regulated transcriptional repression of PCK1 concomitant with the formation of the FOXO1-euchromatic histone-lysine N-methyltransferase2 (EHMT2) complex and histone modifications of the PCK1 promoter region. Notably, our results suggest that FOXO1 uses nucleoporin 98 kDa NUP98 for this transcriptional regulation. These results provide a new insight into various FOXO1-mediated transcriptional regulation and FOXO1-mediated essential biological pathways.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse, clone SC-35, ascites fluid