Merck
Aberrant ATRX protein expression is associated with poor overall survival in NF1-MPNST.
Oncotarget (2018-05-26)
Hsiang-Chih Lu, Vanessa Eulo, Anthony J Apicelli, Melike Pekmezci, Yu Tao, Jingqin Luo, Angela C Hirbe, Sonika Dahiya
ABSTRAKT
Malignant Peripheral Nerve Sheath Tumors (MPNSTs) are aggressive soft tissue sarcomas that can occur sporadically or in the setting of the Neurofibromatosis type 1 (NF1) cancer predisposition syndrome. These tumors carry a dismal overall survival. Previous work in our lab had identified ATRX chromatin remodeler (ATRX), previously termed, Alpha Thalassemia/Mental Retardation Syndrome X Linked as a gene mutated in a subset of MPNSTs. Given the great need for novel biomarkers and therapeutic targets for MPNSTs, we sought to determine the expression of ATRX in a larger subset of sporadic and NF1 associated MPNSTs (NF1-MPNSTs). We performed immunohistochemistry (IHC) on 74 MPNSTs (43 NF1-associated and 31 sporadic), 21 plexiform neurofibromas, and 9 atypical neurofibromas. Using this approach, we have demonstrated that 58% (43/74) of MPNSTs have aberrant ATRX expression (<80% nuclear expression) compared to only 7% (2/30) of benign (plexiform and atypical) neurofibromas. Second, we demonstrated that 65% (28/43) of NF1-MPNSTs displayed aberrant ATRX expression as did 48% (15/31) of sporadic MPNSTs. Finally, we show that aberrant ATRX expression was associated with a significantly decreased overall survival for patients with NF1-MPNST (median OS of 17.9 months for aberrant expression and median OS not met (>120 months) for intact expression, p = 0.0276). In summary, we demonstrate that ATRX is aberrantly expressed in the majority of NF1-MPNSTs, but not plexiform or atypical neurofibromas. Additionally, aberrant ATRX expression is associated with decreased overall survival in NF1-MPNST, but not sporadic MPNST and may serve as a prognostic marker for patients with NF1-MPNST.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-ATRX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution