Merck
Photoinduced Fusion of Lipid Bilayer Membranes.
Langmuir : the ACS journal of surfaces and colloids (2017-02-14)
Yui Suzuki, Ken H Nagai, Anatoly Zinchenko, Tsutomu Hamada
ABSTRAKT
We have developed a novel system for photocontrol of the fusion of lipid vesicles through the use of a photosensitive surfactant containing an azobenzene moiety (AzoTAB). Real-time microscopic observations clarified a change in both the surface area and internal volume of vesicles during fusion. We also determined the optimal cholesterol concentrations and temperature for inducing fusion. The mechanism of fusion can be attributed to a change in membrane tension, which is caused by the solubilization of lipids through the isomerization of AzoTAB. We used a micropipet technique to estimate membrane tension and discuss the mechanism of fusion in terms of membrane elastic energy. The obtained results regarding this novel photoinduced fusion could lead to a better understanding of the mechanism of membrane fusion in living cells and may also see wider applications, such as in drug delivery and biomimetic material design.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Avanti
Cholesterol (ovine), Avanti Research - A Croda Brand