Merck
Telomere-independent cellular senescence in human fetal cardiomyocytes.
Aging cell (2005-01-22)
Andrew J Ball, Fred Levine
ABSTRAKT
Fetal cardiomyocytes have been proposed as a potential source of cell-based therapy for heart failure. This study examined cellular senescence in cultured human fetal ventricular cardiomyocytes (HFCs). HFCs were isolated and identified by immunocytochemistry and RT-PCR. Cells were found to senesce after 20-25 population doublings, as determined by growth arrest, morphological changes and senescence-associated beta-galactosidase activity. Using the telomeric repeat amplification protocol assay, telomerase activity was undetectable in primary HFCs. Cells were transduced to express the human reverse transcriptase subunit (hTERT) of telomerase. This resulted in greatly increased telomerase activity, but no significant lifespan extension. Analysis of telomere length in primary HFCs revealed that the senescent phenotype was not accompanied by telomere shortening. Telomeres in hTERT-positive cells were elongated in comparison with primary cells, and elongation was retained in senescent cells. Levels of the tumor suppressor protein p16INK4A increased in all senescent cells whether telomerase-positive or -negative. Senescence was accompanied by a decline in transcript levels of the polycomb gene Bmi-1, Ets1 and Ets2 transcription factors, and Id1, Id2 and Id3 helix-loop-helix proteins, suggesting roles for these genes in maintenance of cardiomyocyte proliferative capacity. In addition to offering novel insights into the behavior of human fetal cardiomyocytes in culture, these findings have implications for the development of a cell-based therapy for cardiac injury using primary fetal heart tissue.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Cellular Senescence Assay, Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.