- A new C-glycosyl flavone and a new neolignan glycoside from Passiflora edulis Sims peel.
A new C-glycosyl flavone and a new neolignan glycoside from Passiflora edulis Sims peel.
Natural product research (2017-12-05)
Yue Hu, Lin Jiao, Min-Hong Jiang, Sheng Yin, Pan Dong, Zhi-Min Zhao, De-Po Yang, Pei-Tsai Ho, Dong-Mei Wang
PMID29199463
ABSTRAKT
A new C-glycosyl flavone, Chrysin-8-C-(2″-O-β-6-deoxy-glucopyranosyl)-β-D-glucopyranoside (1), a new neolignan glycoside, citrusin G (2), as well as 15 known compounds (3-17) were isolated from the peel of Passiflora edulis Sims. The structure determinations were primarily based on comprehensive spectroscopic analyses, and the absolute configuration of 2 were unequivocally determined by the CD experiment and chemical transformation. Compound 1 represents the rare examples of the flavonoid featuring a deoxy glucose sugar moiety. Compounds 5, 7 and 9 exhibited moderate inhibitory effects on nitric oxide (NO) production stimulated by lipopolysaccharide (LPS) in RAW 264.7 cells, with IC50 values of 34.92, 16.12 and 26.67 μM, respectively.