Merck
T2 mapping in dermatomyositis/polymyositis and correlation with clinical parameters.
Clinical radiology (2018-09-03)
J Ran, S Ji, J N Morelli, G Wu, X Li
ABSTRAKT
To explore the T2-mapping signal characteristics of the thigh muscles in patients with dermatomyositis/polymyositis (DM/PM) and to investigate the correlation between thigh muscle T2 values, clinical parameters, and serum creatinine kinase (CK). Forty-two patients with DM/PM proven by diagnostic criteria were enrolled in the study along with 13 healthy control subjects. Both T2-mapping and conventional magnetic resonance imaging (MRI) images were obtained in the thigh musculature of all subjects. The T2 values of thigh muscles were compared between the DM/PM patients and control groups. Thirty-one DM/PM patients were evaluated with manual muscle testing (MMT) and serum CK levels. A Spearman correlation coefficient model was used to correlate the mean T2 values and clinical assessments. The Kruskal-Wallis test and receiver operating characteristic (ROC) curves were also utilised. p-Values <0.05 reflected statistical significance. The T2 value of all oedematous muscles was greater on average than that of the unaffected muscles of the DM/PM patients (p<0.05) and the muscles of healthy volunteers (p<0.05). The T2 value of unaffected muscles in DM/PM patients was also greater than that of the normal muscles in healthy volunteers (p<0.05). The area under the curves (AUCs) for T2 relaxation time values was 0.72 with respective sensitivity and specificity of 72.6% and 65.4%. The mean T2 relaxation time of the 31 patients group and the MMTs (p<0.05) was correlated without serum CK levels (p>0.05). T2 mapping is not only quantitatively used for subclinical muscle involvement in DM/PM, but also be used to demonstrate severity of damaged muscles in DM/PM.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)